Learn More
BD Prepared Bottled Media: Fluid Thioglycollate (Double Wrapped)
For the detection of microorganisms from pharmaceutical, food, environmental, or water testing samples
Manufacturer:  BD 257264
Catalog No. 22-349-730
Add to cart
Description
Description
Ready for immediate use
Specifications
Specifications
Yellow
Liquid
Yeast extract 5g/L, Pancreatic digest of Casein 15g/L, Glucose 5g/L, L-Cystine 0.5g/L, Sodium Chloride 2.5g/L, Sodium Thioglycollate 0.5g/L, Agar 0.75g/L, Resazurin 0.001g/L
Bottle
5° to 25°C
100mL
United States
Fluid Thioglycollate Medium
10/Pk.
Sterility testing of biologics in sterile filling rooms