Learn More
BD ProbeTec™ Diluent (CT/GC)
For real time DNA amplification assay
Manufacturer:  BD 440453
Catalog No. 22-370-154
This product may not be ordered online. Please contact your sales representative for further ordering instructions.
Includes: Kit contains 4 bottles (225mL a piece)
Description
Description
  • Uses Strand Displacement Amplification (SDA)
  • Used for resuspension of the pelleted specimen before lysis
  • For resuspension of positive and negative controls
  • Batch managed
  • Shelf life: 540 days
Certifications
CE marked
Specifications
Specifications
CT/GC Diluent
4 x 225mL