Learn More
BD Probetec ET CT/GC - Collection Kit for Endocervical Specimens
Used to collect and transport patient endocervical specimens to laboratory for testing
Manufacturer: BD 220142
Alert:
For in vitro diagnostic use
Includes: One large sterile rayon-tipped cleaning swab, one sterile polyurethane-tipped collection swab and one CT/GC diluent tube (2 mL), Diluent composed of potassium phosphate, DMSO, glycerol, Polysorbate 20 and 0.03% Proclin (preservative)
Description
Specimens tested with the BD ProbeTec ET Chlamydia trachomatis and Neisseria gonorrhoeae Amplified DNA Assays
- Consists of sterile collection swab and a CT/GC Diluent tube for specimen transport
- Large rayon-tipped cleaning swab to clean the cervical of blood and mucus prior to collection
- Smaller polyurethane-tipped swab on a plastic shaft to collect the specimen
Specifications
Used to collect and transport patient endocervical specimens to laboratory for testing | |
100 Pack | |
15 to 30°C |
Sterile | |
Plastic | |
One large sterile rayon tipped cleaning swab, one sterile Polyurethane tipped collection swab and one CT/GC diluent tube (2mL), Diluent composed of Potassium Phosphate, DMSO, Glycerol, Polysorbate 20 and 0.03% Proclin™ (preservative) |