Learn More
BD Probetec™ ET CT/GC Collection Kit for Urethral Specimens
Used to collect and transport male patient urethral specimens to laboratory for testing
Manufacturer:  BD 220143
Catalog No. 22-370-160
This product may not be ordered online. Please contact your sales representative for further ordering instructions.
Alert:
For in vitro Diagnostic Use
Includes: Sterile rayon-tipped collection swab, CT/GC diluent tube, Diluent tube (2mL), Diluent composed of potassium phosphate, DMSO, glycerol, Polysorbate 20 and 0.03% Proclin™ (preservative)
Description
Description
The collected specimens are tested with the BD ProbeTec ET Chlamydia trachomatis and Neisseria gonorrhoeae amplified DNA assays
  • Consists of sterile swab and a CT/GC diluent tube for specimen transport
  • Rayon-tipped collection swab with flexible aluminum wire in a plastic shaft
Specifications
Specifications
ET CT or GC Collection Kit
100 Pack
Rayon tip
Used to collect and transport male patient urethral specimens to laboratory for testing
Sterile
Plastic
15 to 30°C
Sterile rayon-tipped collection swab, CT/GC diluent tube, Diluent tube (2mL), Diluent composed of Potassium Phosphate, DMSO, Glycerol, Polysorbate 20 and 0.03% Proclin™ (preservative)