Learn More
BD Probetec™ ET CT/GC Reagent Pack
Use Strand Displacement Amplification (SDA) technology for direct, qualitative detection of Chlamydia trachomatis and Neisseria gonorrhoeae DNA
Manufacturer:  BD 440705
Catalog No. 22-370-152
This product may not be ordered online. Please contact your sales representative for further ordering instructions.
View substitute products »
Description
Description
The BD ProbeTec ET CT/GC assays may be performed using either BD ProbeTec ET System or combination of BD ProbeTec ET System and BD Viper™ instrument.
  • Amplified DNA assays
  • Symptomatic or asymptomatic females and males specimens
  • Tests for presence of C. trachomatis and N. gonorrhoeae DNA in endocervical swabs, male urethral swabs, and in female and male urine specimens
Specifications
Specifications
Chlamydia trachomatis and Neisseria gonorrhoeae DNA
Amplification microwells 4 x 96, restriction Enzyme 30U/15U, DNA Polymerase 25U/2U, dNTP's 80nmol with Buffers and stabilizers
ET CT/GC Reagent Pack
384 Pack