BD Probetec™ ET CT/GC Specimen Collection and Dry Transport System

Offers a convenient way to collect and transport specimens from the patient to the laboratory

Manufacturer: BD 440476

 View more versions of this product

Catalog No. 22-370-167

 N/A
    N/A

This product may not be ordered online. Please contact your sales representative for further ordering instructions. 


    Specifications

    Product Type Endocervical Specimen Collection and Dry Transport Kit
    Closure Color Pink
    For Use With (Equipment) BD Probetec™ ET CT or GC amplified DNA assays
    Includes One Polyurethane-tipped swab in a tube and a large rayon-tipped cleaning swab
    Sterile Yes
    Quantity 100 Pack
    Storage Requirements 15 to 30°C

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit