Learn More
BD Probetec™ ET CT/GC Specimen Collection and Dry Transport System
Offers a convenient way to collect and transport specimens from the patient to the laboratory
Manufacturer:  BD 440461
 View more versions of this product
Catalog No. 22-370-166
This product may not be ordered online. Please contact your sales representative for further ordering instructions.
View substitute products »
Includes: Male Urethral Kit: Each unit includes one rayon-tipped swab on a flexible and extendable wire in a tube
Endocervical Kit: Each unit contains one polyurethane-tipped swab in a tube and a large rayon-tipped cleaning swab
Description
Description
BD ProbeTec™ ET CT/GC Amplified DNA Assay Specimen Collection and DRY TRANSPORT Kit
  • Sterile, self-contained, disposable systems
  • Specialized swabs for collection of urethral or endocervical specimens
  • Swabs facilitate obtaining specimens and maintaining the organisms for nucleic acid amplification
  • For use with the BD ProbeTec ET Chlamydia trachomatis and Neisseria gonorrhoeae Amplified DNA assays on the BD ProbeTec ET System
Endocervical Kit
  • Designed for the cleaning and collection of endocervical specimens
  • Large cleaning swab is included to clean the cervical os of blood and mucus prior to collection
  • Polyurethane-tipped swab on a plastic shaft
Male Urethral Kit
  • Designed for the collection of male urethral specimens
  • Flexible aluminum wire and small rayon-tipped swab facilitate obtaining the specimen
  • Plastic handle housing the aluminum wire and secured in the cap provides a slide action for extension of the swab
Specifications
Specifications
Blue
Sterile
15 to 30°C
One Rayon-tipped swab on a flexible and extendable wire in a tube
Male Urethral Specimen Collection and Dry Transport Kit
100 Pack
BD Probetec™ ET CT or GC amplified DNA assays