Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
BD Probetec™ ET CT Reagent Pack
Uses Strand Displacement Amplification (SDA) technology for direct, qualitative detection of Chlamydia trachomatis DNA
Manufacturer:  BD 440704
Catalog No. 22-370-151
This product may not be ordered online. Please contact your sales representative for further ordering instructions.
Includes: Chlamydia trachomatis (CT) Priming Microwells, 4 x 96: 4 Oligonucleotides ≥7 pmol; dNTP ≥35 nmol; Detector probe ≥25 pmol; with buffers and stabilizers.
Chlamydia trachomatis (CT) Amplification Microwells, 4 x 96: Restriction enzyme ≥30 Units; DNA Polymerase ≥25 Units; dNTP's ≥80 nmol; with buffers and stabilizers.
In addition to the above reagents, the BD ProbeTec ET CT/AC Reagent Pack also contains:
Amplification Control (AC) (Mfr. No. 440474) Priming Microwells, 4 x 96: 4 Oligonucleotides ≥7 pmol; dNTP ≥35 nmol; Detector probe ≥25 pmol; ≥1,000 copies of pGC10 linearized plasmid; with buffers and stabilizers.
Amplification Control (AC) Amplification Microwells, 4 x 96: Restriction enzyme ≥15 Units; DNA Polymerase ≥2 Units; dNTP's ≥80 nmol; with buffers and stabilizers.
This item is not returnable. View return policy
Description
Description
The BD ProbeTec ET CT assay may be performed using either the BD ProbeTec ET System or a combination of the BD ProbeTec ET System and BD Viper™ instrument.
  • Amplified DNA assay
  • Test for presence of C. trachomatis in endocervical swabs, male urethral swabs, and female and male urine specimens as evidence of infection with C. trachomatis.
Specifications
Specifications
Chlamydia trachomatis DNA
Amplification microwells 4 x 96, restriction Enzyme 30U, DNA Polymerase 25U, dNTP's 80nmol, with Buffers and stabilizers
384 Pack
ET CT Reagent Pack