Learn More
BD Probetec ET Instrument
Detection of Chlamydia trachomatis (CT), Neisseria gonorrhoeae (GC), mycobacteria (TB), and atypical pneumonia (AP)
Manufacturer: BD 440478
Description
Real-time DNA amplification assay based on the proprietary BD technology
- Minimal labour and high throughput
- Ease-of-use in a compact, one-room setup
- Performs simultaneous amplification and detection
- One-hour assay format
- Strand Displacement Amplification takes place in sealed amplification microwells to prevent amplicon contamination
- Performs upto 96 reactions simultaneously (including positive and negative controls)
- Results reported as positive, negative, or indeterminate
- Daily maintenance is less than five minutes for the complete system
- LIS connectivity and barcode accession support save time and eliminate errors
Specifications
Chlamydia trachomatis, Neisseria gonorrhoeae, Mycobacteria and Atypical pneumonia | |
ET-instrument | |
96 Reactions |