Learn More
BD Probetec™ Urine Preservative Transport Kit
Designed to preserve and transport urine specimens from symptomatic and asymptomatic individuals
Manufacturer:  BD 440928
Catalog No. 22-370-161
This product may not be ordered online. Please contact your sales representative for further ordering instructions.
Alert:
For in vitro Diagnostic Use
Includes: 100 UPT tubes, 100 transfer pipettes, 50µL of NAP Guard (≥742.5 mM K2EDTA)
Description
Description
  • With NAP Guard™ technology
  • To preserve and transport Chlamydia trachomatis and Neisseria gonorrhoeae in patient's urine specimens prior to processing for analysis with the BD ProbeTec ET CT/GC Reagents
Specifications
Specifications
Urine Preservative Transport Kit
2 to 33°C
100 UPT tubes, 100 transfer pipettes, 50μL of NAP Guard™ (<742.5mM K2EDTA)
100 Pack
Designed to preserve and transport Chlamydia trachomatis and Neisseria gonorrhoeae