Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Protein A: Agarose

Acts as solid matrix facilitating immunoprecipitation of antigen-antibody complexes

Manufacturer:  BD 610437

Catalog No. BDB610437


Add to cart

Description

Description

Protein A has been reported to bind strongly to rabbit IgG and mouse IgG2 isotypes.

  • Storage buffer: Aqueous buffered solution containing ≤0.09% sodium azide

Immunoprecipitation, Western Bloting

Specifications

Specifications

Immunoprecipitation, Western Blotting
Agarose Protein A
4°C
Agarose
1mL
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

Research Use Only