Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Rat Kidney Lysate

Convenient to use and useful control for western blotting application

Manufacturer:  BD 611466

Catalog No. BDB611466


Add to cart

Description

Description

  • Source is rich in endogenously expressed proteins
  • Provided in denatured format allowing for immediate loading onto SDS-PAGE gel
  • Storage buffer: SDS-PAGE buffer (62mM Tris pH 6.8, 2% SDS, 0.9% b-mercaptoethanol, 0.003% bromophenol blue, 5% glycerol)

Routinely tested western blot

Specifications

Specifications

Kidney
1mg/mL in SDS-PAGE Buffer: 62mM Tris pH 6.8, 2% SDS, 0.9% beta-mercaptoethanol, 0.003% Bromophenol Blue, 5% Glycerol
Rat
500μg
-20°C
1mg/mL
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

Research use only