BD Stain Droppers

Identify microorganisms without mixing or measuring stains

Manufacturer: BD 261206

 View more versions of this product

Catalog No. 14-910-77

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Specifications

    Shelf Life 1 to 3 Years
    Storage Requirements 15° to 30°C
    Type Flagella Stain Dropper
    Format Dropper
    For Use With (Application) Used for demonstrating bacterial flagella and their arrangement on the cell
    Quantity 50 Pack
    Certifications/Compliance CE

    To perform tests, squeeze the dropper to crush the ampule. Invert the dropper for convenient drop-by-drop dispensing.

    • Dropper contains one-day supply of reagent or stain, hermetically sealed inside an ampule
    • Strict quality controls assure uniformity and accuracy

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit