BD Nitrate C Reagent Dropper

Identify microorganisms without mixing or measuring reagents

Manufacturer: BD 261207

Catalog No. 14-910-78

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Specifications

    Shelf Life 1 to 3 Years
    Storage Requirements 15° to 30°C
    Stain Type Nitrate Reagent C
    Type Nitrate C Reagent Dropper
    Quantity 50 Pack
    Certifications/Compliance CE
    Format Dropper
    For Use With (Application) For use in determining the ability of bacteria to reduce nitrate to nitrite or free nitrogen gas

    To perform tests, squeeze the dropper to crush the ampule. Invert the dropper for convenient drop-by-drop dispensing.

    • Dropper contains one-day supply of reagent or stain, hermetically sealed inside an ampule
    • Strict quality controls assure uniformity and accuracy

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit