Learn More
BD Reusable Yale Tuberculin Syringes
Reusable syringes with ground barrels and plungers, matched for precision
Catalog Number | Mfr. No. | Volume (Metric) | Graduations | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Graduations | Includes | Price | Quantity & Availability | |||
14-820-15B
|
BD
512002 |
0.5 mL | 0.01 mL | With glass luer-tip |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. Due to product restrictions, please Sign In to purchase or view availability for this product. |
N/A |
14-820-15A
|
BD
512001 |
0.25 mL | 0.01 mL | With glass luer-tip |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. Due to product restrictions, please Sign In to purchase or view availability for this product. |
N/A |
14-820-16
|
BD
512027/DEL |
1 mL | 0.01 mL | Precision with glass luer-tip |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. Due to product restrictions, please Sign In to purchase or view availability for this product. |
N/A |
14-826-36
|
BD
512019 |
2 mL | 0.05 mL | With glass luer-tip |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. Due to product restrictions, please Sign In to purchase or view availability for this product. |
N/A |
14-820-15C
|
BD
512004 |
1 mL | 0.01 mL | With glass luer-tip |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. Due to product restrictions, please Sign In to purchase or view availability for this product. |
N/A |
Description
- Annealed glass permits sterilization by autoclaving
- 2mL syringe (Cat. No. 14-826-36) has dual scale
Specifications
Glass | |
Non-sterile |