Learn More
BD Sedi-Stain™ Concentrated Stain for Urinary Sediment Available on GSA/VA Contract for Federal Government customers only.
No mixing of solutions is required
Manufacturer:  BD 421571
 View more versions of this product
Catalog No. 02-659
Add to cart
Description
Description
  • Stabilized modification of the Sternheimer-Mablin urinary stain
  • Highly selective formula stains blood cells, casts, and other formed elements in urine sediment in a distinctive fashion that permits rapid and accurate identification
  • Sedi-Stain is used directly from the dropper bottle and will remain stable indefinitely
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Highly selective formula stains Blood cells, casts and other formed elements in Urine sediment in a distinctive fashion that permits rapid and accurate identification
60mL
Concentrated stain
Documents
Accessories