Learn More
BD Select Prepared Plated Media Tryptic Soy Broth Agar
For isolation, quantitation and partial identification of many fastidious and non-fastidious microorganisms
Manufacturer:  BD 215269
Catalog No. 22-350-780
Add to cart
This item is not returnable. View return policy
Specifications
Specifications
ISO/DIS, 11014
Liquid
Tryptic Soy broth Agar
10,000mL
Sterile
Light Beige
United States
Plate
2-8°C