Learn More
BD Transduction Laboratories Mouse Kidney Lysate
Rich source of endogenously expressed proteins
Manufacturer:  BD 611457
Catalog No. BDB611457
Add to cart
Alert:
Do not dilute or boil
Description
Description
  • For use as a control for western blotting
  • Provided in a denatured format
  • Ready to use for immediate loading onto an SDS-PAGE gel
Specifications
Specifications
Kidney
1mg/mL in SDS-PAGE Buffer: 62mM Tris pH 6.8, 2% SDS, 0.9% beta-mercaptoethanol, 0.003% Bromophenol Blue, 5% Glycerol
Mouse
500μg
-20°C
1mg/mL