Learn More
BD Transduction Laboratories Mouse Liver Lysate
For use as a control for western blotting
Manufacturer:  BD 611458
Catalog No. BDB611458
Add to cart
Alert:
Do not dilute or boil
Description
Description
  • This lysate can be a rich source of endogenously expressed proteins
  • Provided in a denatured format
  • Ready to use for immediate loading onto an SDS-PAGE gel
Specifications
Specifications
Liver
SDS-PAGE Gel
1 mg/mL
500 μg
Mouse