Learn More
BD Universal Viral Transport Kit Accessories
For use with BD Universal Viral Transport Kits (cat. no. B220527)
Supplier: BD 220239
This item is not returnable.
View return policy
Description
Specifications
Viral Transport Swab | |
Regular tip, sterile, with scored plastic shaft (2/pouch) | |
Plastic |
Collection and transport of clinical specimens containing Viruses, Chlamydiae, Mycoplasmas or Ureaplasmas from the collection site to the testing laboratory | |
100 Pk. | |
Polyester |