Learn More
BD Universal Viral Transport Kit Accessories Available on GSA/VA Contract for Federal Government customers only.
For use with BD Universal Viral Transport Kits (cat. no. B220527)
Manufacturer:  BD 220240
 View more versions of this product
Catalog No. 22-031-18
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
Viral Transport Swab
Plastic
1 Minitip, 1 Regular Tip (1 ea./pouch of 2)
Pk. of 100
Collection and transport of clinical specimens containing Viruses, Chlamydiae, Mycoplasmas or Ureaplasmas from the collection site to the testing laboratory
Polyester