Learn More
BD Vacutainer™ Cornwall™ Metal Syringe Holder
Reusable and sterilizable via EtO or autoclave
Manufacturer:  BD 513527
Catalog No. 22-221655
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Metal holder component of Cornwall continuous pipetter
  • For continuous syringe pipetting
  • Holds 5mL syringes
  • Stainless steel
Specifications
Specifications
Autoclavable
Cornwall™ non-sterile metal syringe
Stainless steel