Learn More
BD Vacutainer™ Glass Blood Collection Tubes with Acid Citrate Dextrose (ACD)
Evacuated glass blood collection tubes for blood banking studies, HLA phenotyping and DNA and paternity testing
Manufacturer:  BD 364012
 View more versions of this product
Catalog No. 02-688-77
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
Blood banking
ACD Solution B, 0.4mL
10.25 x 64mm