BD Vacutainer™ Glass Blood Collection Tubes: Buffered Sodium Citrate

Evacuated glass blood collection tubes for routine coagulation studies

Manufacturer: BD 369714

 View more versions of this product

Catalog No. 02-688-26

  null
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Product Type Coagulation tubes
    Applications Blood Coagulation
    Additive Sodium citrate (0.105M, 3.2%)
    Color Blue
    Length (Metric) Tube 75mm
    Label Type Paper
    Diameter (Metric) Outer 13mm
    Capacity (Metric) 4.5mL
    Closure Type Hemogard

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit