Learn More
BD Vacutainer™ Glass Blood Collection Tubes: Citrate Solution
Evacuated glass blood collection tubes for routine coagulation studies
Manufacturer:  BD 366392
 View more versions of this product
Catalog No. 02-685-25A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • BD Vacutainer Citrate™ tubes with 3.2% buffered sodium citrate solution are used for routine coagulation studies
  • Blood is drawn directly from the vein into the evacuated sterile collection tubes
  • Only one venipuncture needed to draw multiple samples
  • Tubes can be changed while needle and plastic holder remain in place on patient's arm
  • Closures are color coded
Hemogard Closures
  • Safety shield designed for safer tube handling
  • Rubber stopper is recessed inside the plastic shield, so any drops left by a blood collection needle remain isolated from potential contact
  • Shield is designed to protect lab personnel from direct contact with blood on the stopper or around the outer rim of the tube, as well as from blood splattering upon opening the tube
Specifications
Specifications
1.8mL draw
Sodium citrate (0.105M, 3.2%)
47mm
10.25mm
Conventional
Blue
Paper