Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Vacutainer™ Glass Blood Collection Tubes with SPS for Microbiology Testing

Evacuated blood culture specimen collection tubes for whole blood microbiology testing

Specifications

Applications Microbiology Testing
Color Yellow
Material Glass
Label Type Paper
View More Specs

Products 1
Catalog Number Mfr. No. Diameter (Metric) Outer Capacity (Metric) Length (Metric) Tube Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Outer Capacity (Metric) Length (Metric) Tube Price Quantity & Availability  
02-684-19
View Documents
BD
364960
8.3mL 8.3 mL Draw 100mm
Pack of 100 for $134.33
Case of 10 PK for $934.19
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

  • Blood is drawn directly from the vein into the evacuated sterile collection tubes
  • Only one venipuncture is needed to draw multiple samples
  • Tubes can be changed while needle and plastic holder remain in place on patient's arm
  • Yellow color-coded stoppers to enhance speed and accuracy in the laboratory

Specifications

Specifications

Microbiology Testing
Yellow
Glass
Paper
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit