Learn More
BD Vacutainer™ Glass Blood Collection Tubes with SPS for Microbiology Testing
Evacuated blood culture specimen collection tubes for whole blood microbiology testing
Manufacturer:  BD 366404
 View more versions of this product
Catalog No. 02-684-20
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Blood is drawn directly from the vein into the evacuated sterile collection tubes
  • Only one venipuncture is needed to draw multiple samples
  • Tubes can be changed while needle and plastic holder remain in place on patient's arm
  • Yellow color-coded stoppers to enhance speed and accuracy in the laboratory
Specifications
Specifications
Microbiology Testing
Yellow
Paper
Glass
62mm
10.25mm