Learn More
BD Vacutainer™ Glass Blood Collection Tubes for Westergren Sed Rate Determination
Evacuated glass blood collection tubes for Westergren sedimentation rate (ESR) determination
Manufacturer:  BD 369741
 View more versions of this product
Catalog No. 02-688-36
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • Blood is drawn directly from the vein into the evacuated sterile collection tubes
  • Only one venipuncture is needed to draw multiple samples
  • Tubes can be changed while needle and plastic holder remain in place on patient's arm
  • Closures are color coded
Hemogard Closures
  • Safety shield designed for safer tube handling
  • Rubber stopper is recessed inside the plastic shield, so any drops left by a blood collection needle remain isolated from potential contact
  • Shield is designed to protect lab personnel from direct contact with blood on the stopper or around the outer rim of the tube, as well as from blood splattering upon opening the tube
Specifications
Specifications
For Westergren ESR determinations
13 x 75mm; Draw: 2mL
Glass