BD Vacutainer™ Plastic Blood Collection Tubes with K2EDTA: Tube Stopper

Evacuated blood collection tubes for whole blood hematology determinations

Manufacturer: BD 368661

 View more versions of this product

Catalog No. 23-021-013

  null
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Material Plastic
    Additive K2EDTA, 10.8mg
    Color Lavender
    Length (Metric) Tube 100mm
    Diameter (Metric) Outer 13mm
    Capacity (Metric) 6mL
    Closure Type Conventional

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit