Learn More
BD Vacutainer™ Urine Culture and Sensitivity (C&S) Transport Kits
Contain a freeze-dried preservative for transporting and preserving urine specimens
Products 1
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
14-375-131
View Documents
BD
364949
N/A Draw: 5mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • The kit comes with a BD Vacutainer tube and a transfer straw that delivers urine from the cup into the evacuated tube
  • Preservative prevents overgrowth of bacteria, even if the specimen is contaminated