Learn More
BD Vacutainer™ Venous Blood Collection Tubes: Vacutainer Plus™ Glass Serum Tubes, Silicone-Coated, with Conventional Stopper
Use for serum determinations in chemistry, serology and immunohematology
Manufacturer:  BD 366431
 View more versions of this product
Catalog No. 02-685B
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Sterile vacuum blood collection tubes for serum determination
  • Spray-coated silica interior
  • With conventional tube stopper
Evacuated Tubes Simplify Blood Collection
  • Blood is drawn directly from the vein into the evacuated sterile collection tubes
  • Only one venipuncture is needed to draw multiple samples
  • Tubes can be changed while needle and plastic holder remain in place on patient's arm
  • Closures are color coded
Specifications
Specifications
13 times x 100mm; 7.0mL
Glass
Red
Conventional
Silicone coated