Learn More
BD Red Nucleic Acid Stain
Useful for discrimination of viable from nonviable cells, DNA content analysis in fixed cells, or nuclear counterstaining for fixed cells in immunofluorescence or flow cytometry applications. BD Via-Probe™ Red Nucleic Acid Stain brightly stains nonviable or fixed cells with permeable membranes.
Manufacturer:  BD 565804
 View more versions of this product
Catalog No. BDB565804
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Red Nucleic Acid Stain has an excitation maximum of 631nm, and is best excited by the red laser
  • It has an emission maximum of 651nm
Specifications
Specifications
0.5 mL
  • Immunofluorescence
  • Immunocytochemistry
  • Flow cytometry
  • Intracellular staining (flow cytometry)
Red
Red Nucleic Acid Stain