Learn More
BD Yale™ Reusable Hypodermic Needles
Fit either BD Luer-Lok™ or standard Luer-tip syringes
Manufacturer:  BD 511109
 View more versions of this product
Catalog No. 22-222281
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Fit either BD Luer-Lok™ or standard Luer-tip syringes
  • Corrosion-resistant stainless-steel cannula is double swaged to hub, providing strong needle assembly with unrestricted bore
  • Specially contoured hub interior eliminates hard-to-clean crevices and permits easy needle wire insertion
  • Four-point locking flange
Specifications
Specifications
13 G
Four-point locking flange
2 in.
Metal
Non-sterile
Luer-lock or standard luer-tip syringes
Yes
50.8 mm
Regular Bevel
Yale Hypodermic Needle