Learn More
BD Yale Reusable Hypodermic Needles
Fit either BD Luer-Lok or standard Luer-tip syringes
Specifications
For Use With (Equipment) | Luer-lock or standard luer-tip syringes |
---|---|
Includes | Four-point locking flange |
Material (Hub) | Metal |
Type | Yale Hypodermic Needle |
Catalog Number | Mfr. No. | Needle Gauge | Length (English) Needle | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Needle Gauge | Length (English) Needle | Price | Quantity & Availability | ||||
22-222281
|
BD
511109 |
13 G | 2 in. |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. Due to product restrictions, please Sign In to purchase or view availability for this product. |
N/A |
Description
Fit either BD Luer-Lok or standard Luer-tip syringes
- Corrosion-resistant stainless-steel cannula is double swaged to hub, providing strong needle assembly with unrestricted bore
- Specially contoured hub interior eliminates hard-to-clean crevices and permits easy needle wire insertion
- Four-point locking flange
Specifications
Luer-lock or standard luer-tip syringes | |
Four-point locking flange | |
Metal | |
Yale Hypodermic Needle |