BD Yale™ Reusable Hypodermic Needles
Fit either BD Luer-Lok™ or standard Luer-tip syringes
Manufacturer: BD 511055
 View more versions of this product
Catalog No. 14-826-27
    N/A
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 
    Specifications
    Gauge 22 ga
    Length (English) Needle 1 in.
    Length (Metric) Needle 25.4mm
    Type Yale™ Hypodermic Needle
    For Use With (Equipment) Luer-lock or standard luer-tip syringes
    Includes Four-point locking flange
    Material (Hub) Metal
    Needle Point Style Regular Bevel
  • Description & Specifications
Related Products
Product Content Feedback