Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BDCS, silylation reagent, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass Bottle
1185092-02-7
C6H15ClSi·C3H4N2
218.8
MFCD00000501
IFLPAOFWVBWJPG-UHFFFAOYSA-N
bdcs silylation reagent,bdcs, silylation reagent, acroseal,him; t-butyldimethylchlorosilane,tert-butyldimethylchlorosilane imidazole,t-butyl-dimethylsilyl chloride-imidazole,imidazole; t-butyldimethylchlorosilane,tert-butyl-chloro-dimethylsilane; 1h-imidazole
2724131
tert-butyl-chloro-dimethylsilane;1H-imidazole
CC(C)(C)[Si](C)(C)Cl.C1=CN=CN1
1185092-02-7
C6H15ClSi·C3H4N2
218.8
MFCD00000501
IFLPAOFWVBWJPG-UHFFFAOYSA-N
bdcs silylation reagent,bdcs, silylation reagent, acroseal,him; t-butyldimethylchlorosilane,tert-butyldimethylchlorosilane imidazole,t-butyl-dimethylsilyl chloride-imidazole,imidazole; t-butyldimethylchlorosilane,tert-butyl-chloro-dimethylsilane; 1h-imidazole
2724131
tert-butyl-chloro-dimethylsilane;1H-imidazole
CC(C)(C)[Si](C)(C)Cl.C1=CN=CN1
Color Yellow
Flash Point 58°C
Density 0.9590g/mL
Specific Gravity 0.959
Packaging AcroSeal™ Glass Bottle
Refractive Index 1.4400 to 1.4420
Quantity 800mL
Infrared Spectrum Authentic
Fieser 07,184; 01,489; 03,157; 08,255
Formula Weight 218.8
Physical Form Solution
Chemical Name or Material BDCS
Show More Show Less

GHS H Statement
Flammable liquid and vapour.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Suspected of damaging the unborn child.
Contact with water liberates toxic gas.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta

GHS Signal Word: Danger