Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BDH2 Mouse anti-Human, Clone: OTI4G4, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525141

Catalog No. PIMA525141


Add to cart

Description

Description

The function of this protein remains unknown.
Specifications

Specifications

BDH2
Monoclonal
0.73 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9BUT1
BDH2; DHRS6; 3-hydroxybutyrate dehydrogenase, type 2; EFA6R; FLJ13261; PRO20933; UCPA-OR; UNQ6308; dehydrogenase/reductase (SDR family) member 6; oxidoreductase UCPA
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
56898
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI4G4
Unconjugated
BDH2
Liquid
BDH2
Full length human recombit protein of BDH2 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit