Learn More
BDNF, Guinea Pig anti-Human,Mouse,Rat,Monkey, Polyclonal, Novus Biologicals™
Guinea Pig Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP280493
Catalog No. NB013566
Add to cart
Description
Description
BDNF Polyclonal antibody specifically detects BDNF in Human, Mouse, Rat, Monkey Samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications
Specifications
BDNF
Unconjugated
Lyophilized from whole serum
abrineurin, brain-derived neurotrophic factor, MGC34632, neurotrophin
Guinea Pig
Unpurified
RUO
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Polyclonal
Human, Monkey, Mouse, Rat
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot 1:500, Immunohistochemistry 1:500, Immunohistochemistry-Paraffin 1:500
Unpurified
BDNF
A synthetic peptide from c-terminal region of mature mouse BDNF conjugated to blue carrier protein was used as the antigen.
0.1 mL
Cardiovascular Biology, Cytokine Research, Immunology, Neuroscience
Primary
627