Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BDNF Mouse anti-Human, Biotin, Clone: 3D9C5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237276B

Catalog No. NBP237276B

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

BDNF
3D9C5
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Purified
BDNF
Purified recombinant fragment of human BDNF (AA: 19-248) expressed in E. Coli.
Protein A or G purified
Cardiovascular Biology, Cytokine Research, Immunology, Neuroscience
Primary
627
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Biotin
PBS with 0.05% Sodium Azide
abrineurin, brain-derived neurotrophic factor, MGC34632, neurotrophin
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit