Learn More
Gibco™ BDNF Recombinant Human Protein Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.
BDNF Recombinant Human Protein
Manufacturer:  Gibco™ 10908010
 View more versions of this product
Catalog No. 10-908-010
Add to cart
Description
Description
Recombinant Human Brain-Derived Neurotrophic Factor (BDNF) is a bioactive protein intended for use in cell culture applications. BDNF is a neurotrophic factor involved in the survival and differentiation of neurons in the central and peripheral nervous system and used in the maintenance of motor neurons and the differentiation of both cholinergic and dopaminergic neurons.
  • High purity—no interference from other proteins or contaminants
  • High biological activity—more results with less protein
  • Proven compatibility—Gibco™ proteins bioassayed with Gibco™ media
High Purity Recombinant Bioactive Protein
Recombinant Human Brain-Derived Neurotrophic Factor is one of our more than 250 available Gibco™ recombinant proteins. To help ensure Gibco™ growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein.
High Biological Activity
In-house activity testing of Gibco™ Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, antiviral, and protease assays. Recombinant Human Brain-Derived Neurotrophic Factor (BDNF) specific activity is determined by measuring the dose-dependent induction of choline acetyl transferase activity in primary septal cell cultures.
Proven Compatibility
Gibco™ proteins are bioassayed with Gibco™ media. Since 1962, Gibco™ has been the consistent provider of high quality media, reagents, and sera for reliable cell culture.
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
BDNF Recombinant Human Protein, 5μg, Lyophilized Form, <0.1ng/μg Endotoxin Level, –5 to –30°C Storage, 96 % Purity, For 3D Cell Culture (Extracellular Matrices), Cell Viability, Proliferation and Function, Mammalian Cell Culture,
Cell culture, mammalian cell culture, 3D cell culture (extracellular matrices), cell analysis, cell viability, proliferation and function, primary cell culture, neuronal cell culture, stem cell research, stem cell culture, stem cell differentiation, embry
Lyophilized
BDNF Recombinant Human Protein
5μg, 5μg
–5 to –30°C
<0.1ng/μg
Lyophilized
1 Each
96%
RUO - research use only
–5 to –30°C
Documents
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only. Not for use in diagnostic procedures.