Learn More
Gibco™ BDNF Recombinant Human Protein Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.
$341.00 - $402.00
Products 2
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
10-908-010 Non-distribution product as customer accommodation.
View Documents
Gibco™
10908010
N/A 5μg
Each for $341.00
Add to cart
PHC7074 Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.
View Documents
Gibco™
PHC7074
N/A 10μg
Each for $402.00
Add to cart
Description
Description
Recombinant Human Brain-Derived Neurotrophic Factor (BDNF) is a bioactive protein intended for use in cell culture applications. BDNF is a neurotrophic factor involved in the survival and differentiation of neurons in the central and peripheral nervous system and used in the maintenance of motor neurons and the differentiation of both cholinergic and dopaminergic neurons.
  • High purity—no interference from other proteins or contaminants
  • High biological activity—more results with less protein
  • Proven compatibility—Gibco™ proteins bioassayed with Gibco™ media
High Purity Recombinant Bioactive Protein
Recombinant Human Brain-Derived Neurotrophic Factor is one of our more than 250 available Gibco™ recombinant proteins. To help ensure Gibco™ growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein.
High Biological Activity
In-house activity testing of Gibco™ Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, antiviral, and protease assays. Recombinant Human Brain-Derived Neurotrophic Factor (BDNF) specific activity is determined by measuring the dose-dependent induction of choline acetyl transferase activity in primary septal cell cultures.
Proven Compatibility
Gibco™ proteins are bioassayed with Gibco™ media.
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.