Learn More
Chemglass Life Sciences 20L Bead Bath, 230V, Supplied with 16L of Bath Beads
SDP
Supplier:  Chemglass Life Sciences CLS4598009E
Encompass_Preferred
20L Bead Bath, 230V, Supplied with 16L of Bath Beads. Bead bath gets up and running faster and you never need to turn it off. There is nothing to evaporate. It’s just like a trusty, always-on incubator. Unlike a standard bead-filled, water bath, Bead Bath allows you to achieve a new set temperature quickly, without long warm up delays, so you can make quick adjustments on the fly.With Bead Bath designed specifically for beads, temperatures are consistent and uniform everywhere in the bath. Thermal uniformity is an amazing ±0.5°C at 37°C and ±1.0 at 65°C.
Catalog No. 50-143-562