Learn More
Fisherbrand™ Beaker Heater and PID Control Bundle with J-type Thermocouple
Fisherbrand™ Beaker Heater and PID Control Bundle with J-type Thermocouple features a silicone rubber beaker heater, ideal for heating standard size glass or plastic beakers.
 View more versions of this product
Catalog No. 11-476-163
Add to cart
Description
Description
  • Beaker Heater
  • Easy-to-install and remove with hook and loop fastener
  • Moisture and chemical resistant
  • Uniform heat across surface of beaker
  • Hook and loop closure allows for quick installation and removal
  • Compact, plug-and-play design
  • Advanced PID control
  • Simple three-key user control
  • Type J thermocouple sensor input
Warranty and Services
1-year
Specifications
Specifications
±0.9°F (0.50°C)
Digital PID Benchtop Controller
76 mm
120 V
J-type thermocouple
Heater and Control Bundle
Heating glass or plastic beakers
PID
6.39 x 9.48 x 1.99 in.
311 mm
Beaker Heater, Digital PID Temperature Controller
150 W
232°C