Learn More
Zyagen LabsSupplier Diversity Partner BEANS GENOMIC DNA: Intact highly pure DNA extracted from plant leaves, treated with DNase & packed at a concentration of 0.5mg/ml or 1mg/ml in TE buffer. Ideal for PCR, library construction & many other applications DFS Item
Manufacturer:  Zyagen LabsSupplier Diversity Partner PLG1051
Zyagen tissue genomic DNA is intact highly pure extracted from plant leaves, treated with DNase-free RNase to remove contaminant RNA, precisely measured by Nanodrop and packed in 1.5-ml tubes at a concentration of 0.5mg/ml or 1mg/ml in TE buffer. Applications: PCR Amplification, Southern blot analysis, Genomic DNA methylation, Single-nucleotide polymorphism (SNP) Analysis, Profiling studies in gene expression, and DNA library construction. Quality Control: The integrity and purity of genomic DNA was tested by agarose gel electrophoresis and Nanodrop, Ratio of A260/280 is >1.8, Free of RNA contamination, Successful PCR amplification of Beta-Actin, and Successful digestion of DNA by EcoR1 and/or Hind III enzyme. 
Catalog No. 50-171-9209