Learn More
Great Lakes Diagnostics™ Beckman™ Compatible Filtered Tips
Sterile, filtered tips are compatible with Beckman™ Robotic Systems
Supplier: Great Lakes Diagnostics™ K2504800
Description
- Compatible Beckman Coulter™ systems include:
- NF
- NX
- I5
- I7
Specifications
250 μL | |
Sterile | |
Autoclavable | |
960 Tips/Cs. | |
Single-use |
Filtered | |
Colorless | |
Hanging Tip Rack | |
Natural | |
No |