Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HemoCue America Beckman Coulter™ Hemoccult™ ICT Immunochemical Fecal Occult Blood Test Kit
Supplier: HemoCue America 395068A
Includes: ICT FOBT Kit (Cat. No. 23-280-012) includes test devices and 8mL bottle of Hemoccult ICT Buffer; ICT Sample Collection Kit (Cat. No. 23-280-011) contains Patient Kits; each Patient Kit contains Dispensing Envelope with Patient Instructions, Hemoccult ICT Collection Cards, Collection Tissues, Applicator Sticks, and Mailing Pouch; ICT Collection Cards (Cat. No. 23-280-010) contain Collection Cards and Applicator Sticks; ICT Control Kit (Cat. No. 23-280-013) contains bottles (Positive and Negative)
Description
- CLIA Complexity: Waived
- CPT Codes: 82274QW, G0328QW
- Rapid, qualitative immunochemical chromatographic method for detection of human hemoglobin from blood in fecal samples
- Visually read results
Specifications
82274QW, G0328QW | |
ICT Control Kit | |
Bottles (positive and negative) |
Waived | |
Detection of Human hemoglobin from Blood in fecal samples | |
4 Pack |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction