Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Beclomethasone dipropionate, ≥99%, MilliporeSigma™ Supelco™
Beclomethasone dipropionate is a type of steroid drug, administered by means of inhalation. It can behave like an anti-inflammatory agent within the respiratory tract. It can also decrease airway resistance and reduce mucosal oedema.
Supplier: MilliporeSigma Supelco B30225G
Specifications
5534-09-8 | |
Analytical Standard | |
MFCD00135613 | |
521.04 | |
Analytical Standard | |
Neat |
5 g | |
C28H37 ClO7 | |
9-Chloro-11 beta,17,21-trihydroxy-16 beta-methylpregna-1,4-diene-3,20-dione 17,21-dipropionate | |
≥99% | |
Drug Analysis |
Safety and Handling
Recommended Storage : 2°C to 8°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction