Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beclomethasone Dipropionate, Micronized, USP, 97-103%, Spectrum™

Click to view available options
Quantity:
250 mg
1 g
5 g
100 g
1 kg
Packaging:
Amber Glass Bottle
5534-09-8
C28H37ClO7
521.05
KUVIULQEHSCUHY-XYWKZLDCSA-N
2-[(1R,2S,3aS,3bS,9aS,9bR,10S,11aS)-9b-chloro-10-hydroxy-2,9a,11a-trimethyl-7-oxo-1-(propanoyloxy)-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]-2-oxoethyl propanoate
CCC(=O)OCC(=O)[C@@]1(OC(=O)CC)[C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]12C
5534-09-8
C28H37ClO7
521.05
KUVIULQEHSCUHY-XYWKZLDCSA-N
2-[(1R,2S,3aS,3bS,9aS,9bR,10S,11aS)-9b-chloro-10-hydroxy-2,9a,11a-trimethyl-7-oxo-1-(propanoyloxy)-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]-2-oxoethyl propanoate
CCC(=O)OCC(=O)[C@@]1(OC(=O)CC)[C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]12C

B1412, 5534-09-8, C28H37ClO7

Spectrum™ Beclomethasone Dipropionate, Micronized, USP is potent glucocorticoid steroid used to prevent and control symptoms caused by asthma. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Ignition Residue 0.1%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Specific Rotation +88° to +94°
Quantity 1 g
CAS Max % 100%
Percent Purity 97 to 103%