Learn More
BECN1 Mouse anti-Human, Clone: OTI4A10, liquid, TrueMAB™
SDP
Mouse Monoclonal Antibody
Supplier:  OriGene TA502617
Encompass_Preferred
Catalog No. 50-167-3657
Add to Cart
Description
Description
Beclin 1 plays a central role in autophagy. It acts as a core subunit of the PI3K complex that mediates formation of phosphatidylinositol 3-phosphate. Different PI3K complex forms are believed to play a role in multiple membrane trafficking pathways - PI3KC3-C1 is involved in the initation of autophagosomes and PI3KC3-C2 is involved in the maturation of autophagosomes and endocytosis. Beclin 1 also plays a role in antiviral host defense.
Specifications
Specifications
BECN1
Monoclonal
Unconjugated
BECN1
ATG6, VPS30, beclin1
Mouse
Affinity Chromatography
RUO
8678
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI4A10
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q14457
BECN1
Full length human recombit protein of human BECN1 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG2a
Product Suggestions
Product Suggestions