Learn More
Sklar InstrumentsSupplier Diversity Partner Beeson Plaster Spreader, Sklar, 12 in., 2 Flat Blades Outside Serrated, Straight, Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 111812
Beeson Plaster Spreader, Sklar, 12 in., 2 Flat Blades Outside Serrated, Straight, Stainless Steel. Sklar's Beeson Plaster Spreader is used to aid in the removal of plaster casts. It has a solid straight handle and two large, flat, rectangular blades with horizontal serrations on the outside of the blade. Once a cut is made in the plaster cast, the blades of the spreader are closed by opening the handle and are inserted to a section of the cast. By squeezing the handle, the blades spread open, there by spreading apart sections of the cast. After multiple sections of the cast are spread open, the cast may be removed. The blades are approximately 2-1/8 inches wide and the overall length of this instrument is 12 inches.
Catalog No. 50-143-7329